Gene Transcripts

Gene Symbol
PDCD1
Entrez Gene ID
5133
Gene Name
programmed cell death 1
Gene Aliases
CD279, PD-1, PD1, SLEB2, hPD-1, hPD-l, hSLE1
Chromosome Location
Chr. 2: 241849881 - 241858908 on Build GRCh38
UniGene ID
Hs.158297
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005018.2 NP_005009.2 2 379
XM_006712573.2 XP_006712636.1 2 381
XM_017004293.1 XP_016859782.1 2 381
GenBank mRNA AK313848.1 - 2 381
AY206416.1 - 1 141
AY238517.1 - 2 311
BC074740.2 - 2 357 30915198
KJ865859.1 - 2 311
KJ865860.1 - 2 311
KJ865861.1 - 2 311
U64863.1 - 2 379

View Full Product Details