Gene Transcripts

Gene Symbol
PTPRCAP
Entrez Gene ID
5790
Gene Name
protein tyrosine phosphatase, receptor type C associated protein
Gene Aliases
CD45-AP, LPAP
Chromosome Location
Chr. 11: 67435510 - 67437682 on Build GRCh38
UniGene ID
Hs.155975
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005608.2 NP_005599.1 2 183
GenBank mRNA AK312022.1 - 2 183
BC113859.1 - 2 182 40030438
BC114467.1 - 2 182 40030437
CR457429.1 - 1 120
X81422.1 - 2 183

View Full Product Details