Gene Transcripts

Gene Symbol
SPP1
Entrez Gene ID
6696
Gene Name
secreted phosphoprotein 1
Gene Aliases
BNSP, BSPI, ETA-1, OPN
Chromosome Location
Chr. 4: 87975650 - 87983411 on Build GRCh38
UniGene ID
Hs.313
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000582.2 NP_000573.1 6 874
NM_001040058.1 NP_001035147.1 7 916
NM_001040060.1 NP_001035149.1 6 835
NM_001251829.1 NP_001238758.1 5 793
NM_001251830.1 NP_001238759.1 8 1105
XM_017008564.1 XP_016864053.1 5 1342
GenBank mRNA AF052124.1 - 6 796
AK290090.1 - 6 771
AK290104.1 - 7 853
AK295491.1 - 8 1040
AK296035.1 - 6 771
AK315461.1 - 6 804
BC007016.1 - 6 810 4052438
BC017387.1 - 7 856 3828885
BC022844.1 - 6 811 4284921
BC093033.1 - 6 827 5288027
BX648003.1 - 6 789
DQ839491.1 - 6 751
DQ846870.1 - 6 751
J04765.1 - 6 817
JF412667.1 - 6 833
KR062183.1 - 8 1040
X13694.1 - 7 818

View Full Product Details