Gene Transcripts

Gene Symbol
SDC3
Entrez Gene ID
9672
Gene Name
syndecan 3
Gene Aliases
SDCN, SYND3
Chromosome Location
Chr. 1: 30869466 - 30909735 on Build GRCh38
UniGene ID
Hs.158287
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014654.3 NP_055469.3 3 649
XM_011542463.1 XP_011540765.1 3 584
XM_011542464.2 XP_011540766.1 3 854
XM_011542466.1 XP_011540768.1 3 491
GenBank mRNA AB007937.1 - 1 1931
AF248634.1 - 3 656
AK295698.1 - 2 394
KU178484.1 - 2 386
KU178485.1 - 1 428

View Full Product Details