Gene Transcripts

Gene Symbol
RAMP1
Entrez Gene ID
10267
Gene Name
receptor activity modifying protein 1
Gene Aliases
Chromosome Location
Chr. 2: 237858714 - 237912117 on Build GRCh38
UniGene ID
Hs.471783
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308353.1 NP_001295282.1 4 620
NM_005855.3 NP_005846.1 3 559
XM_017003152.1 XP_016858641.1 3 481
XM_017003153.1 XP_016858642.1 3 789
XM_017003154.1 XP_016858643.1 3 566
XM_017003156.1 XP_016858645.1 3 507
GenBank mRNA AJ001014.1 - 3 459
AY265457.1 - 3 427
BC000548.2 - 3 451 3163522
BI548986.1 - 4 546
BM126103.1 - 1 360
BM978161.1 - 1 363
CR542032.1 - 3 427
CR542044.1 - 3 427

View Full Product Details