Gene Transcripts

Gene Symbol
ERLIN2
Entrez Gene ID
11160
Gene Name
ER lipid raft associated 2
Gene Aliases
C8orf2, Erlin-2, NET32, SPFH2, SPG18
Chromosome Location
Chr. 8: 37736579 - 37757801 on Build GRCh38
UniGene ID
Hs.705490
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003790.3 NP_001003790.1 4, 5 583
NM_001003791.2 NP_001003791.1 4, 5 354
NM_007175.6 NP_009106.1 4, 5 354
XM_005273392.2 XP_005273449.1 4, 5 514
XM_017013000.1 XP_016868489.1 4, 5 352
GenBank mRNA AK291394.1 - 4, 5 307
AK297279.1 - 4, 5 307
AL442077.1 - 341
AY358108.1 - 4, 5 298
AY358851.1 - 4, 5 288
BC005950.1 - 4, 5 277
BC048308.1 - 4, 5 577
BC050611.1 - 4, 5 308
BC067765.1 - 4, 5 581 5296776
BI560439.1 - 4, 5 587
BP353279.1 - 4, 5 354

View Full Product Details