Gene Transcripts

Gene Symbol
EFR3B
Entrez Gene ID
22979
Gene Name
EFR3 homolog B
Gene Aliases
KIAA0953
Chromosome Location
Chr. 2: 25042104 - 25159137 on Build GRCh38
UniGene ID
Hs.4892
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319099.1 NP_001306028.1 16 1951
NM_014971.1 NP_055786.1 16 1963
XM_011532701.2 XP_011531003.1 16 1936
GenBank mRNA AB023170.1 - 16 1937
AK095190.1 - 16 1951
BC049384.1 - 13 1507 5742159

View Full Product Details