Gene Transcripts

Gene Symbol
PDCD11
Entrez Gene ID
22984
Gene Name
programmed cell death 11
Gene Aliases
ALG-4, ALG4, NFBP, RRP5
Chromosome Location
Chr. 10: 103396615 - 103446295 on Build GRCh38
UniGene ID
Hs.239499
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014976.1 NP_055791.1 22 3542
XM_005269647.3 XP_005269704.1 22 3582
XM_011539538.2 XP_011537840.1 22 3530
XM_011539539.2 XP_011537841.1 22 3530
XM_011539540.1 XP_011537842.1 21 3396
GenBank mRNA BC049838.1 - 22 3543
BC064486.1 - 22 3535
BC080560.1 - 22 3535
BC111040.1 - 22 3497
D80007.1 - 22 3495

View Full Product Details