Gene Transcripts
- Gene Symbol
- PHF24
- Entrez Gene ID
- 23349
- Gene Name
- PHD finger protein 24
- Gene Aliases
- KIAA1045
- Chromosome Location
- Chr. 9: 34810040 - 34982544 on Build GRCh38
- UniGene ID
- Hs.7989
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001304333.2 |
NP_001291262.1
|
7, 8 |
1311 |
|
NM_015297.2 |
NP_056112.1
|
7, 8 |
1307 |
|
NR_136307.1 |
|
7, 8 |
1311 |
|
NR_136308.1 |
|
7, 8 |
1307 |
|
XM_011517827.1 |
XP_011516129.1
|
8, 9 |
1299 |
|
XM_011517828.2 |
XP_011516130.1
|
7, 8 |
1403 |
|
XM_011517829.1 |
XP_011516131.1
|
7, 8 |
1561 |
|
XM_017014553.1 |
XP_016870042.1
|
9, 10 |
1593 |
|
XM_017014554.1 |
XP_016870043.1
|
9, 10 |
1373 |
|
XM_017014555.1 |
XP_016870044.1
|
9, 10 |
1345 |
|
XM_017014556.1 |
XP_016870045.1
|
9, 10 |
1877 |
|
GenBank mRNA |
AB028968.1 |
- |
7, 8 |
1178 |
|
AK294342.1 |
- |
7, 8 |
1311 |
|
View Full Product Details