Gene Transcripts

Gene Symbol
NAT9
Entrez Gene ID
26151
Gene Name
N-acetyltransferase 9 (putative)
Gene Aliases
EBSP, hNATL
Chromosome Location
Chr. 17: 74770529 - 74776425 on Build GRCh38
UniGene ID
Hs.144058
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001305077.1 NP_001292006.1 6 634
NM_001305078.1 NP_001292007.1 6 633
NM_001305080.1 NP_001292009.1 6 622
NM_001305081.1 NP_001292010.1 6 631
NM_001305082.1 NP_001292011.1 6 782
NM_001305083.1 NP_001292012.1 6 619
NM_015654.4 NP_056469.2 6 637
NR_130953.1 7 641
XM_005257212.4 XP_005257269.1 5 476
XM_006721813.3 XP_006721876.1 5 924
XM_017024437.1 XP_016879926.1 6 563
GenBank mRNA AK123115.1 - 5 850
AK225602.1 - 6 507
AK308569.1 - 6 915
AK312958.1 - 6 507
AL050269.1 - 6 514
AL537925.3 - 6 714
AY632082.1 - 5 473
BC004195.2 - 6 543 3528894
BC004225.2 - 6 532 3538186
BI821280.1 - 7 538
BM909550.1 - 6 521
BP231776.1 - 6 555
BU159218.1 - 6 529

View Full Product Details