Gene Transcripts

Gene Symbol
TSPAN17
Entrez Gene ID
26262
Gene Name
tetraspanin 17
Gene Aliases
FBX23, FBXO23, TM4SF17
Chromosome Location
Chr. 5: 176647387 - 176659058 on Build GRCh38
UniGene ID
Hs.532129
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006616.2 NP_001006617.2 3 499
NM_012171.2 NP_036303.1 3 499
NM_130465.4 NP_569732.2 3 499
GenBank mRNA AF174603.1 - 3 309
AK024427.1 - 3 389
BC010405.2 - 3 321 3621871
BC067105.1 - 3 384 6646576

View Full Product Details