Gene Transcripts

Gene Symbol
CECR1
Entrez Gene ID
51816
Gene Name
cat eye syndrome chromosome region, candidate 1
Gene Aliases
ADA2, ADGF, IDGFL, PAN, SNEDS
Chromosome Location
Chr. 22: 17178790 - 17221854 on Build GRCh38
UniGene ID
Hs.170310
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282225.1 NP_001269154.1 8 1497
NM_001282226.1 NP_001269155.1 8 1464
NM_001282227.1 NP_001269156.1 8 1270
NM_001282228.1 NP_001269157.1 8 1372
NM_001282229.1 NP_001269158.1 7 1129
NM_177405.2 NP_803124.1 5 703
XM_006724080.3 XP_006724143.1 7 2734
XM_011546133.1 XP_011544435.1 8 1389
GenBank mRNA AB209648.1 - 7 2734
AF190746.1 - 7 1433
AK027682.1 - 5 524
AK074702.1 - 5 575
AK130725.1 - 2 217
AK292689.1 - 8 1485
AK304818.1 - 8 1264
AK314321.1 - 8 1288
BC051755.1 - 7 1268 5753141
CR456417.1 - 7 1255
DA864284.1 - 5 570

View Full Product Details