Gene Transcripts

Gene Symbol
PRR13
Entrez Gene ID
54458
Gene Name
proline rich 13
Gene Aliases
TXR1
Chromosome Location
Chr. 12: 53441649 - 53446643 on Build GRCh38
UniGene ID
Hs.744894
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005354.2 NP_001005354.1 3 297
NM_018457.3 NP_060927.1 3 447
GenBank mRNA AF217517.1 - 1 360
AK074398.1 - 1 343
BC001464.2 - 3 433
BC004512.2 - 1 40
BC014257.2 - 1 347 4765821
BC016064.1 - 1 344 4852862
BC021019.1 - 1 315
BC066943.1 - 1 346 4817871
BC071979.1 - 1 290
BX640962.1 - 2 185
CN392914.1 - 3 461
CR533459.1 - 2 283

View Full Product Details