Gene Transcripts

Gene Symbol
PNMAL2
Entrez Gene ID
57469
Gene Name
paraneoplastic Ma antigen family-like 2
Gene Aliases
Chromosome Location
Chr. 19: 46491191 - 46495912 on Build GRCh38
UniGene ID
Hs.7193
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020709.2 NP_065760.1 1 1699
GenBank mRNA AB033009.1 - 1 376
BC121810.1 - 1 333
BC122520.1 - 1 333

View Full Product Details