Gene Transcripts

Gene Symbol
PCDH19
Entrez Gene ID
57526
Gene Name
protocadherin 19
Gene Aliases
EFMR, EIEE9
Chromosome Location
Chr. X: 100291644 - 100410273 on Build GRCh38
UniGene ID
Hs.4993
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105243.1 NP_001098713.1 4 4245
NM_001184880.1 NP_001171809.1 5 4386
NM_020766.2 NP_065817.2 4 4242
XM_011530997.2 XP_011529299.1 5 3352
GenBank mRNA AB037734.1 - 4 460
AK096591.1 - 4 1198
BC136628.1 - 4 1126 9020618
CR749278.1 - 4 1123
EF676096.1 - 5 4386

View Full Product Details