Gene Transcripts
- Gene Symbol
- ANAPC1
- Entrez Gene ID
- 64682
- Gene Name
- anaphase promoting complex subunit 1
- Gene Aliases
- APC1, MCPR, TSG24
- Chromosome Location
- Chr. 2: 111766147 - 111884196 on Build GRCh38
- UniGene ID
- Hs.436527
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022662.3 |
NP_073153.1
|
18, 19 |
2361 |
|
XM_011511633.2 |
XP_011509935.1
|
18, 19 |
2393 |
|
XM_011511634.2 |
XP_011509936.1
|
18, 19 |
2508 |
|
XM_011511636.2 |
XP_011509938.1
|
18, 19 |
2393 |
|
XM_011511637.2 |
XP_011509939.1
|
18, 19 |
2393 |
|
XM_017004710.1 |
XP_016860199.1
|
18, 19 |
2393 |
|
XM_017004711.1 |
XP_016860200.1
|
18, 19 |
2393 |
|
XM_017004712.1 |
XP_016860201.1
|
18, 19 |
2508 |
|
XM_017004713.1 |
XP_016860202.1
|
17, 18 |
2456 |
|
XM_017004714.1 |
XP_016860203.1
|
17, 18 |
2404 |
|
XM_017004715.1 |
XP_016860204.1
|
18, 19 |
2393 |
|
GenBank mRNA |
AJ278357.1 |
- |
18, 19 |
2376 |
|
AK023807.1 |
- |
5, 6 |
587 |
|
AL832013.1 |
- |
7, 8 |
717 |
|
BC104902.1 |
- |
18, 19 |
2163 |
8143905 |
BC104904.1 |
- |
18, 19 |
2163 |
8143907 |
- Gene Symbol
- LOC102724642
- Entrez Gene ID
- 102724642
- Gene Name
- anaphase-promoting complex subunit 1-like
- Gene Aliases
- Chromosome Location
- Chr. 2: 87125147 - 87171376 on Build GRCh38
- Species
- Homo sapiens
View Full Product Details