Gene Transcripts

Gene Symbol
CENPO
Entrez Gene ID
79172
Gene Name
centromere protein O
Gene Aliases
CENP-O, ICEN-36, MCM21R
Chromosome Location
Chr. 2: 24793115 - 24822376 on Build GRCh38
UniGene ID
Hs.731569
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199803.2 NP_001186732.1 2, 3 558
NM_001322101.1 NP_001309030.1 3, 4 672
NM_024322.3 NP_077298.1 3, 4 691
NR_136182.1 2, 3 577
NR_136183.1 2, 3 577
NR_136184.1 2, 3 558
GenBank mRNA AK027859.1 - 2, 3 292
AK315483.1 - 3, 4 317
AL514456.3 - 2, 3 251
BC002870.1 - 3, 4 348 3942844
BP366489.1 - 3, 4 344
BX406097.2 - 3, 4 347
CN263048.1 - 2, 3 214
CX872478.1 - 2, 3 195

View Full Product Details