Gene Transcripts

Gene Symbol
ASXL3
Entrez Gene ID
80816
Gene Name
additional sex combs like 3, transcriptional regulator
Gene Aliases
BRPS, KIAA1713
Chromosome Location
Chr. 18: 33578577 - 33751195 on Build GRCh38
UniGene ID
Hs.464876
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030632.2 NP_085135.1 12 4015
XM_005258356.1 XP_005258413.1 13 3988
XM_011526205.2 XP_011524507.1 13 10778
XM_011526206.2 XP_011524508.1 13 7543
XM_011526209.1 XP_011524511.1 14 4172
XM_011526210.1 XP_011524512.1 14 4229
XM_011526212.1 XP_011524514.1 13 4133
XM_011526213.1 XP_011524515.1 13 4226
XM_017026012.1 XP_016881501.1 13 7390
XM_017026013.1 XP_016881502.1 14 7752
XM_017026014.1 XP_016881503.1 14 4189
GenBank mRNA AB051500.2 - 2 2174

View Full Product Details