Gene Transcripts

Gene Symbol
THAP3
Entrez Gene ID
90326
Gene Name
THAP domain containing 3
Gene Aliases
Chromosome Location
Chr. 1: 6624645 - 6635586 on Build GRCh38
UniGene ID
Hs.689006
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195752.1 NP_001182681.1 1, 2 132
NM_001195753.1 NP_001182682.1 2, 3 222
NM_138350.3 NP_612359.2 1, 2 132
XM_005263532.3 XP_005263589.1 2, 3 442
XM_011542401.2 XP_011540703.1 1, 2 90
XM_017002761.1 XP_016858250.1 1, 2 90
GenBank mRNA AK096217.1 - 1, 2 90
BC092427.1 - 2, 3 222 30376814
BE905510.1 - 191

View Full Product Details