Gene Transcripts

Gene Symbol
DDX60L
Entrez Gene ID
91351
Gene Name
DEAD-box helicase 60-like
Gene Aliases
Chromosome Location
Chr. 4: 168356735 - 168480514 on Build GRCh38
UniGene ID
Hs.535011
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012967.2 NP_001012985.2 25 3563
NM_001291510.1 NP_001278439.1 25 3563
XM_005263341.4 XP_005263398.2 25 3546
XM_005263342.2 XP_005263399.2 25 3563
XM_005263343.2 XP_005263400.2 25 3529
XM_005263345.4 XP_005263402.2 25 3542
XM_006714408.2 XP_006714471.1 26 3613
XM_011532402.2 XP_011530704.1 26 3836
XM_011532403.2 XP_011530705.1 26 3671
XM_011532404.1 XP_011530706.1 24 3467
XM_011532405.1 XP_011530707.1 24 3467
XM_011532406.1 XP_011530708.1 25 3563
XM_011532409.2 XP_011530711.1 13 1603
XM_017008829.1 XP_016864318.1 24 3433
XM_017008830.1 XP_016864319.1 25 3563
XM_017008831.1 XP_016864320.1 14 1865
GenBank mRNA AK023743.1 - 4 448
AK055595.1 - 16 2062
AK092461.1 - 18 2403
AL832950.1 - 6 658
CR933601.1 - 25 3538
KR632542.1 - 23 3315

View Full Product Details