Gene Transcripts

Gene Symbol
TTYH2
Entrez Gene ID
94015
Gene Name
tweety family member 2
Gene Aliases
C17orf29
Chromosome Location
Chr. 17: 74213557 - 74262020 on Build GRCh38
UniGene ID
Hs.27935
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032646.5 NP_116035.5 12, 13 1477
NM_052869.1 NP_443101.1 6, 7 1287
XM_005257824.3 XP_005257881.1 12, 13 1405
GenBank mRNA AF319952.1 - 12, 13 1456
AK058197.1 - 1, 2 177
AK091085.1 - 10, 11 1158
AK126955.1 - 6, 7 1695
AK296511.1 - 12, 13 1400
AK315163.1 - 12, 13 1463
AL831858.1 - 2, 3 223
BC005168.1 - 12, 13 1452
BC107492.1 - 12, 13 1473 40033786
BC107602.1 - 12, 13 1473 40033791
BX647912.1 - 6, 7 1287

View Full Product Details