Gene Transcripts

Gene Symbol
TTYH2
Entrez Gene ID
94015
Gene Name
tweety family member 2
Gene Aliases
C17orf29
Chromosome Location
Chr. 17: 74213557 - 74262020 on Build GRCh38
UniGene ID
Hs.27935
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032646.5 NP_116035.5 7 870
NM_052869.1 NP_443101.1 1 680
XM_005257824.3 XP_005257881.1 7 798
GenBank mRNA AF319952.1 - 7 849
AK091085.1 - 5 551
AK126955.1 - 1 1088
AK296511.1 - 7 793
AK315163.1 - 7 856
BC005168.1 - 7 845
BC107492.1 - 7 866 40033786
BC107602.1 - 7 866 40033791
BX647912.1 - 1 680

View Full Product Details