Gene Transcripts

Gene Symbol
WFIKKN1
Entrez Gene ID
117166
Gene Name
WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 1
Gene Aliases
C16orf12, RJD2, WFDC20A, WFIKKN
Chromosome Location
Chr. 16: 631012 - 634116 on Build GRCh38
UniGene ID
Hs.345818
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053284.2 NP_444514.1 2 1656
GenBank mRNA AF422194.1 - 2 1414
AK075356.1 - 2 1735
AK128777.1 - 1 4584
AK314757.1 - 2 1718
BC101602.1 - 2 1497 8069108
BC101606.1 - 2 1497 8069112

View Full Product Details