Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136200.1 | NP_001129672.1 | 5 | 1613 | |
NM_144591.3 | NP_653192.2 | 6 | 816 | ||
GenBank mRNA | AK098071.1 | - | 5 | 1562 | |
AL832477.1 | - | 5 | 1602 | ||
BC015994.1 | - | 6 | 768 | 4698381 |