Gene Transcripts

Gene Symbol
HYKK
Entrez Gene ID
123688
Gene Name
hydroxylysine kinase
Gene Aliases
AGPHD1
Chromosome Location
Chr. 15: 78507564 - 78537373 on Build GRCh38
UniGene ID
Hs.307962
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013619.3 NP_001013641.2 2 141
NM_001083612.1 NP_001077081.1 2 141
XM_011521231.2 XP_011519533.1 2 123
XM_017021922.1 XP_016877411.1 2 123
GenBank mRNA AK126453.1 - 2 123
BC132753.1 - 2 88 40146775
BC144383.1 - 2 88 9052904
BM045979.1 - 2 142

View Full Product Details