Gene Transcripts

Gene Symbol
C7orf13
Entrez Gene ID
129790
Gene Name
chromosome 7 open reading frame 13
Gene Aliases
MY040
Chromosome Location
Chr. 7: 156638366 - 156640654 on Build GRCh38
UniGene ID
Hs.647014
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026865.1 1 1073
GenBank mRNA AF063598.1 - 1 840
AF439977.1 - 1 1073
AK056371.1 - 1 989
BC031037.1 - 1 958
BC070228.1 - 2 398

View Full Product Details