Gene Transcripts

Gene Symbol
SNTN
Entrez Gene ID
132203
Gene Name
sentan, cilia apical structure protein
Gene Aliases
S100A1L, S100AL, sentan
Chromosome Location
Chr. 3: 63652668 - 63665217 on Build GRCh38
UniGene ID
Hs.130704
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080537.1 NP_001074006.1 3 268
GenBank mRNA AK126350.1 - 3 238

View Full Product Details