Gene Transcripts

Gene Symbol
C5orf47
Entrez Gene ID
133491
Gene Name
chromosome 5 open reading frame 47
Gene Aliases
Chromosome Location
Chr. 5: 173989159 - 174007524 on Build GRCh38
UniGene ID
Hs.131469
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144954.1 NP_001138426.1 3, 4 612
XM_017009028.1 XP_016864517.1 3, 4 606
GenBank mRNA BC034971.1 - 3, 4 578
BC047225.1 - 3, 4 567
BC053953.1 - 3, 4 567
BC057842.1 - 3, 4 553
BC066964.1 - 3, 4 574
BC092489.1 - 3, 4 601

View Full Product Details