Gene Transcripts

Gene Symbol
TPM3P9
Entrez Gene ID
147804
Gene Name
tropomyosin 3 pseudogene 9
Gene Aliases
Chromosome Location
Chr. 19: 53431974 - 53444672 on Build GRCh38
UniGene ID
Hs.433293
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003148.3 2 1097
GenBank mRNA AK094777.1 - 4 1037
BC061910.1 - 2 908
BX647962.1 - 2 1084

View Full Product Details