Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_003087.1 | 3 | 481 | ||
NR_003088.1 | 3 | 481 | |||
GenBank mRNA | AF418600.1 | - | 3 | 481 | |
AF518320.1 | - | 3 | 250 | ||
AF518321.1 | - | 3 | 336 | ||
AY063514.1 | - | 3 | 361 | ||
AY063515.1 | - | 3 | 400 | ||
AY344117.1 | - | 3 | 389 | ||
BC144077.1 | - | 2 | 214 | ||
DR004370.1 | - | 3 | 492 |