Gene Transcripts

Gene Symbol
GGN
Entrez Gene ID
199720
Gene Name
gametogenetin
Gene Aliases
Chromosome Location
Chr. 19: 38384347 - 38388603 on Build GRCh38
UniGene ID
Hs.447948
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152657.3 NP_689870.3 3 1890
XM_005258619.4 XP_005258676.1 3 2252
XM_011526603.2 XP_011524905.1 3 2002
XM_017026451.1 XP_016881940.1 2 2096
GenBank mRNA AF538035.1 - 3 1895
AF538036.1 - 4 954
AK093032.1 - 2 1838
BC035008.2 - 3 1890 5269088

View Full Product Details