Gene Transcripts
- Gene Symbol
- LOC202181
- Entrez Gene ID
- 202181
- Gene Name
- SUMO interacting motifs containing 1 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 5: 177618500 - 177672277 on Build GRCh38
- UniGene ID
- Hs.189914
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026921.1 |
|
6 |
2039 |
|
GenBank mRNA |
AF086197.1 |
- |
1 |
445 |
|
AK021623.1 |
- |
1 |
422 |
|
View Full Product Details