Gene Transcripts

Gene Symbol
LINC01566
Entrez Gene ID
283914
Gene Name
long intergenic non-protein coding RNA 1566
Gene Aliases
Chromosome Location
Chr. 16: 35363416 - 35391713 on Build GRCh38
UniGene ID
Hs.385772
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027079.2 3 619
NR_027080.2 3 432
GenBank mRNA BC038774.1 - 3 432
BC045579.1 - 3 619

View Full Product Details