Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001123366.1 | NP_001116838.1 | 4 | 459 | |
XM_011525930.2 | XP_011524232.1 | 4 | 554 | ||
XM_017025710.1 | XP_016881199.1 | 4 | 554 | ||
GenBank mRNA | AW205887.1 | - | 1 | 178 | |
BM696260.1 | - | 4 | 459 |