Gene Transcripts

Gene Symbol
C17orf51
Entrez Gene ID
339263
Gene Name
chromosome 17 open reading frame 51
Gene Aliases
Chromosome Location
Chr. 17: 21524606 - 21551678 on Build GRCh38
UniGene ID
Hs.514016
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113434.4 NP_001106905.1 2 782
GenBank mRNA AK023039.1 - 1 90
AK056436.1 - 2 748
BC010612.1 - 2 780
BC019938.1 - 2 770
BC052777.1 - 2 782
BC053952.1 - 2 782
BC132738.1 - 2 756 40146760
BC136630.1 - 2 756 9020620
BX640850.1 - 3, 4 673

View Full Product Details