Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001012643.3 | NP_001012661.1 | 2 | 336 | |
XM_017026742.1 | XP_016882231.1 | 1 | 1081 | ||
XM_017026743.1 | XP_016882232.1 | 1 | 1081 | ||
GenBank mRNA | BC044311.1 | - | 2 | 336 | |
BU196283.1 | - | 228 |