Gene Transcripts
- Gene Symbol
- RBM44
- Entrez Gene ID
- 375316
- Gene Name
- RNA binding motif protein 44
- Gene Aliases
- Chromosome Location
- Chr. 2: 237798048 - 237842808 on Build GRCh38
- UniGene ID
- Hs.607265
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001080504.2 |
NP_001073973.2
|
11, 12 |
2660 |
|
XM_005246075.1 |
XP_005246132.1
|
11, 12 |
3001 |
|
XM_011511155.1 |
XP_011509457.1
|
3, 4 |
169 |
|
XM_017004054.1 |
XP_016859543.1
|
11, 12 |
3004 |
|
XM_017004055.1 |
XP_016859544.1
|
11, 12 |
3374 |
|
XM_017004056.1 |
XP_016859545.1
|
11, 12 |
2944 |
|
XM_017004058.1 |
XP_016859547.1
|
6, 7 |
1342 |
|
GenBank mRNA |
AK130316.1 |
- |
2, 3 |
99 |
|
BC126111.1 |
- |
8, 9 |
743 |
|
BX648107.1 |
- |
10, 11 |
3313 |
|
View Full Product Details