Gene Transcripts

Gene Symbol
LOC101927168
Entrez Gene ID
101927168
Gene Name
uncharacterized LOC101927168
Gene Aliases
Chromosome Location
Chr. 18: 6728927 - 6729862 on Build GRCh38
UniGene ID
Hs.242240
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_134645.1 2 351
GenBank mRNA AW135595.1 - 1 46
BG202214.1 - 1 212
BX097886.1 - 2 351

View Full Product Details