Gene Transcripts
- Gene Symbol
- RBMS3-AS3
- Entrez Gene ID
- 100873979
- Gene Name
- RBMS3 antisense RNA 3
- Gene Aliases
- Chromosome Location
- Chr. 3: 29264194 - 29280939 on Build GRCh38
- UniGene ID
- Hs.99391
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_109804.1 |
|
1, 2 |
207 |
|
GenBank mRNA |
BM840572.1 |
- |
1, 2 |
207 |
|
View Full Product Details