Gene Transcripts
- Gene Symbol
- FLJ22447
- Entrez Gene ID
- 400221
- Gene Name
- uncharacterized LOC400221
- Gene Aliases
- Chromosome Location
- Chr. 14: 61570540 - 61654713 on Build GRCh38
- UniGene ID
- Hs.344000
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_039985.1 |
|
3 |
360 |
|
GenBank mRNA |
AK026100.1 |
- |
3 |
364 |
|
BQ439352.1 |
- |
3 |
453 |
|
View Full Product Details