Gene Transcripts

Gene Symbol
LOC100419583
Entrez Gene ID
100419583
Gene Name
ring finger protein 4 pseudogene
Gene Aliases
Chromosome Location
Chr. 15: 44728988 - 44732851 on Build GRCh38
UniGene ID
Hs.489254
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_125376.1 1 344
GenBank mRNA AK311550.1 - 1 311
BC031266.1 - 1 210
DC350419.1 - 1 344

View Full Product Details