Gene Transcripts
- Gene Symbol
- C1orf143
- Entrez Gene ID
- 440714
- Gene Name
- chromosome 1 open reading frame 143
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.146774
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BG219097.1 |
- |
1, 2 |
96 |
|
View Full Product Details