Gene Transcripts

Gene Symbol
KCTD21
Entrez Gene ID
283219
Gene Name
potassium channel tetramerization domain containing 21
Gene Aliases
KCASH2
Chromosome Location
Chr. 11: 78171243 - 78188854 on Build GRCh38
UniGene ID
Hs.528439
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029859.2 NP_001025030.1 2 368
XM_005273925.3 XP_005273982.1 2 593
XM_005273928.1 XP_005273985.1 3 493
XM_006718516.3 XP_006718579.1 2 3538
XM_006718517.2 XP_006718580.1 3 605
XM_006718518.3 XP_006718581.1 2 353
XM_011544955.2 XP_011543257.1 2 1833
GenBank mRNA AK300320.1 - 2 355
BC036058.1 - 2 357 5277643
DC342183.1 - 2 368

Gene Symbol
KCTD21-AS1
Entrez Gene ID
100289388
Gene Name
KCTD21 antisense RNA 1
Gene Aliases
Chromosome Location
Chr. 11: 78139793 - 78173977 on Build GRCh38
UniGene ID
Hs.628183
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC062331.1 - 1 495

View Full Product Details