Gene Transcripts

Gene Symbol
LOC401357
Entrez Gene ID
401357
Gene Name
uncharacterized LOC401357
Gene Aliases
Chromosome Location
Chr. 7: 56809124 - 56812736 on Build GRCh38
UniGene ID
Hs.457696
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_130727.1 1 889
GenBank mRNA AK125790.1 - 1 888
AK128114.1 - 1 630
BC137417.1 - 1 624
JX508854.1 - 1 599

View Full Product Details