Gene Transcripts
- Gene Symbol
- LOC100130698
- Entrez Gene ID
- 100130698
- Gene Name
- uncharacterized LOC100130698
- Gene Aliases
- Chromosome Location
- Chr. 10: 80247758 - 80255408 on Build GRCh38
- UniGene ID
- Hs.447456
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_136612.1 |
|
6 |
1148 |
|
GenBank mRNA |
AK125824.1 |
- |
1 |
3379 |
|
AK128140.1 |
- |
1 |
1864 |
|
View Full Product Details