Gene Transcripts

Gene Symbol
POLR2F
Entrez Gene ID
5435
Gene Name
RNA polymerase II subunit F
Gene Aliases
HRBP14.4, POLRF, RPABC14.4, RPABC2, RPB14.4, RPB6, RPC15
Chromosome Location
Chr. 22: 37953663 - 38041915 on Build GRCh38
UniGene ID
Hs.436578
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK098727.1 - 1 979
AL832562.1 - 1 1213

View Full Product Details