Gene Transcripts

Gene Symbol
DNAL4
Entrez Gene ID
10126
Gene Name
dynein axonemal light chain 4
Gene Aliases
MRMV3, PIG27
Chromosome Location
Chr. 22: 38778508 - 38794156 on Build GRCh38
UniGene ID
Hs.632766
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005740.2 NP_005731.1 3 362
GenBank mRNA AK223349.1 - 3 349
AK290566.1 - 3 315
AL035366.1 - 3 355
AY453399.1 - 153
BC002968.1 - 3 329 3543962
BT009847.1 - 2 134
BX473356.1 - 3 367
CR456487.1 - 2 219
CR542177.1 - 2 134
CR542195.1 - 2 134

View Full Product Details