Gene Transcripts

Gene Symbol
USPL1
Entrez Gene ID
10208
Gene Name
ubiquitin specific peptidase like 1
Gene Aliases
C13orf22, D13S106E, bA121O19.1
Chromosome Location
Chr. 13: 30617397 - 30660770 on Build GRCh38
UniGene ID
Hs.533831
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321532.1 NP_001308461.1 8 1735
NM_001321533.1 NP_001308462.1 7 1095
NM_001321534.1 NP_001308463.1 6 824
NM_005800.4 NP_005791.3 9 1888
XM_005266213.3 XP_005266270.1 10 2246
XM_006719751.3 XP_006719814.1 9 2166
XM_017020314.1 XP_016875803.1 8 2070
XM_017020315.1 XP_016875804.1 7 1441
XM_017020316.1 XP_016875805.1 9 2079
GenBank mRNA AK057514.1 - 6 824
AK289826.1 - 9 1644
BC036027.1 - 9 1660 5271146
BC038103.2 - 9 1798 4821753
CR627370.1 - 7 838
X59131.1 - 9 1633

View Full Product Details