Gene Transcripts

Gene Symbol
TIMM17B
Entrez Gene ID
10245
Gene Name
translocase of inner mitochondrial membrane 17 homolog B (yeast)
Gene Aliases
DXS9822, JM3, TIM17B
Chromosome Location
Chr. X: 48893447 - 48898143 on Build GRCh38
UniGene ID
Hs.30570
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167947.2 NP_001161419.1 5 447
NM_005834.4 NP_005825.1 4 297
GenBank mRNA AF034790.1 - 4 297
AF077039.1 - 3 162
AJ005895.1 - 3 150
AK290207.1 - 3 172
BC010142.2 - 3 151 4300787
CB993524.1 - 4 338

View Full Product Details