Gene Transcripts

Gene Symbol
TMC6
Entrez Gene ID
11322
Gene Name
transmembrane channel like 6
Gene Aliases
EV1, EVER1, EVIN1, LAK-4P
Chromosome Location
Chr. 17: 78108775 - 78132427 on Build GRCh38
UniGene ID
Hs.632227
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127198.2 NP_001120670.1 6 767
NM_001321185.1 NP_001308114.1 6 720
NM_007267.7 NP_009198.4 6 671
XM_005256995.1 XP_005257052.1 6 796
XM_011524255.1 XP_011522557.1 5 698
XM_011524256.1 XP_011522558.1 5 698
XM_011524257.2 XP_011522559.1 1 39
XM_011524258.1 XP_011522560.1 5 698
XM_017024107.1 XP_016879596.1 6 745
XM_017024108.1 XP_016879597.1 6 698
XM_017024109.1 XP_016879598.1 5 584
GenBank mRNA AK021738.1 - 5 625
AK074065.1 - 6 670
AK096076.1 - 6 703
AK303165.1 - 2 244
AK310734.1 - 5 1091
AY057379.1 - 5 510
AY099356.1 - 7 669
AY099357.1 - 7 669
AY236496.1 - 5 636
BC007766.1 - 5 536
BC023597.2 - 6 651 4566166

View Full Product Details